Learn More
Medchemexpress LLC C12H12N4O2S2 | 675104-49-1 | 99.8% | 308.38 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY113794100MG
Encompass_Preferred
DSHS00884 is a potent human papillomavirus E6 inhibitor with an IC50 of 10 μM. It stabilizes p53 levels in HPV-positive cells by blocking E6-mediated p53 degradation with an IC50 of 2.5-25 μM.
  • Potent human papillomavirus E6 inhibitor
  • Ic50 of 10 μM against human papillomavirus E6
  • Stabilizes p53 levels in HPV-positive cells
  • Blocks E6-mediated p53 degradation
  • Ic50 of 2.5-25 μM for blocking E6-mediated p53 degradation
Catalog No. 50-004-36361