Learn More
Aviva Systems Biology Corp C12orf24 Antibody - N-terminal region (ARP54929_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54929_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C12orf24. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C12orf24. Alternate names: HSU79274, C12orf24ApplicationWB. Species reactivity: Cow, Human, Pig
Catalog No. 50-014-61371