Learn More
Medchemexpress LLC C14-4 | 2639634-80-1 | 98.3% | 1365.30 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY148842
Encompass_Preferred
C14-4 is an ionizable lipid utilized for the synthesis of lipid nanoparticles (LNPs). It enhances mRNA delivery, enabling the effective transport of mRNA to primary human T cells, which in turn induces functional protein expression. C14-4 demonstrates high transfection efficiency while maintaining low cytotoxicity.
  • Utilized for lipid nanoparticle (LNP) synthesis.
  • Enhances mRNA delivery to primary human T cells.
  • Induces functional protein expression in human T cells.
  • Demonstrates high transfection efficiency.
  • Maintains low cytotoxicity.
  • Promotes luciferase expression in Jurkat cells via mRNA delivery.
Catalog No. 50-003-63275