Learn More
Medchemexpress LLC C14-4 | 2639634-80-1 | 99.0% | 1365.30 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1488421MG
Encompass_Preferred
C14-4 is an ionizable lipid used in the creation of lipid nanoparticles (LNPs). It is designed to enhance mRNA delivery, specifically enabling the efficient transport of mRNA to primary human T cells, which subsequently leads to functional protein expression. This compound exhibits high transfection efficiency while maintaining low cytotoxicity.
  • Utilized for the synthesis of lipid nanoparticles (LNPs).
  • Enhances mRNA delivery to primary human T cells.
  • Induces functional protein expression.
  • Demonstrates high transfection efficiency.
  • Maintains low cytotoxicity.
Catalog No. 50-003-62366