Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC C14-4 | 2639634-80-1 | 99.01% | 1365.30 | 1 ML
SDP

Supplier:  Medchemexpress LLC HY14884210MM/1ML

Encompass_Preferred

C14-4 is an ionizable lipid utilized for the synthesis of lipid nanoparticles (LNPs). It enhances mRNA delivery, enabling the effective transport of mRNA to primary human T cells, which in turn induces functional protein expression. C14-4 demonstrates high transfection efficiency while maintaining low cytotoxicity.

  • Used for synthesis of lipid nanoparticles (LNPs)
  • Enhances mRNA delivery to primary human T cells
  • Induces functional protein expression
  • Exhibits high transfection efficiency
  • Maintains low cytotoxicity
  • Appearance: liquid
  • Color: colorless to light yellow
  • Pure form storage: -20°C for 3 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month
  • Solubility in ethanol: 100 mg/mL
  • Solubility in DMSO: 100 mg/mL

Catalog No. 50-003-60861


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.