Learn More
Medchemexpress LLC C14-4 | 2639634-80-1 | 99.01% | 1365.30 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1488425MG
Encompass_Preferred
C14-4 is an ionizable lipid used for the synthesis of lipid nanoparticles (LNPs). It enhances mRNA delivery, facilitating the effective transport of mRNA to primary human T cells, which then induces functional protein expression.
  • Enhances mRNA delivery
  • Facilitates transport to primary human T cells
  • Induces functional protein expression
  • Shows high transfection efficiency
  • Exhibits low cytotoxicity
Catalog No. 50-003-62149