Learn More
Cayman Chemical C15 GalactosylcRamIDd18 1 5mg
SDP
Supplier:  Cayman Chemical 244665 MG
Encompass_Preferred
A glycosphingolipid; contains a galactose moiety attached to a ceramide acylated with pentadecanoic acid
Catalog No. 50-293-2579