Learn More
Medchemexpress LLC C15H9Cl2NO2 | 79183-19-0 | 98.6% | 306.14 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1863310MM/1ML
Encompass_Preferred
MDK83190 is a potent apoptosis activator that induces Apaf-1 oligomerization, increases procaspase-9 processing, and subsequently activates caspase-3 in a cytochrome c-dependent manner. It is intended for research use only and not for sale to patients.
  • Potent apoptosis activator
  • Induces Apaf-1 oligomerization
  • Increases procaspase-9 processing
  • Activates caspase-3 in a cyto c-dependent manner
Catalog No. 50-003-87737