Learn More
Cayman Chemical C16 MPEG 2000 CeramIded1810mg
SDP
Supplier:  Cayman Chemical 4195510 MG
Encompass_Preferred
A PEGylated form of C16 ceramide; has been used in the generation of microspheres, LNPs, and liposomes for the delivery of small molecules, mRNA, and plasmid DNA in vitro and in vivo; liposomes containing C16 ceramide-mPEG(2000) with a stearylated-INF7 and have been used to transfect plasmid DNA into MFLM-4 cells; microspheres containing C16 ceramide-mPEG(2000) and encapsulating oxaliplatin increase survival and decrease the number of small intestine, colon, and cecum polyps in an Apc+/Minz spontaneous colorectal adenoma mouse model
Catalog No. 50-294-3698