Learn More
Medchemexpress LLC C16-PAF | 74389-68-7 | 99.86% | 523.68 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10863510MG
Encompass_Preferred
C16-PAF (PAF (C16)), a phospholipid mediator, is a platelet-activating factor and ligand for PAF G-protein-coupled receptor (PAFR). C16-PAF exhibits anti-apoptotic effect and inhibits caspase-dependent death by activating the PAFR. C16-PAF is a potent MAPK and MEK/ERK activator. C16-PAF induces increased vascular permeability.
  • Phospholipid mediator.
  • Platelet-activating factor and ligand for PAF G-protein-coupled receptor (PAFR).
  • Exhibits anti-apoptotic effect and inhibits caspase-dependent death by activating the PAFR.
  • Potent MAPK and MEK/ERK activator.
  • Induces increased vascular permeability.
  • For research use only.
Catalog No. 50-003-46587