Learn More
Medchemexpress LLC C16 PEG2000 ceramide | 212116-78-4 | MFCD02265697 | 99.4% | 2000 (Average) | (C2H4O)nC39H73NO6 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1440055MG
Encompass_Preferred
C16 PEG2000 ceramide is a polyethylene glycol-conjugated C16 ceramide (average PEG molecular weight 2000) used as a lipid component in delivery formulations and laboratory research. It is reported to induce autophagy and is commonly applied in studies of lipid-based carriers and cancer-related cellular assays.
  • PEGylated C16 ceramide with average PEG molecular weight 2000.
  • Functions as a lipid carrier for formulation and delivery studies.
  • Reported to induce autophagy in cellular assays.
  • High chemical purity suitable for research applications.
  • Available in small milligram pack sizes for laboratory use.
Catalog No. 50-002-21207