Learn More
Cayman Chemical C17 SphngomyelInd18 117 0 5mg
SDP
Supplier:  Cayman Chemical 255925 MG
Encompass_Preferred
A synthetic derivative of sphingomyelin; has been used as an internal standard for the quantification of sphingomyelin
Catalog No. 50-293-4321