Learn More
Avanti Polar Lipids C18:0 trisialoganglioside-GT1b (d18:1/18:0, triammonium salt) 100µg
Supplier:  Avanti Polar Lipids 860089W100UG
Encompass
C18:0 GT1b is a trisialoganglioside. It belongs to the group of gangliosides, composed of glycosphingolipids and three sialic acids.Gangliosides occur predominantly on the outer surface of cell membranes and, together with glycoproteins and glycosaminoglycans, it forms the glycocalyx that covers eukaryotic cell surfaces.The role of GT1b-Trisialogangliosides is mainly associated with the nervous system. They are located especially in the axon membranes, where the molecules bind to myelin-associated glycoproteins. This interaction is crucial to regulating and maintaining axon-myelin stability.
Catalog No. NC3735137