Learn More
Cayman Chemical C18 D-threo CeramIded18 11 1mg
SDP
Supplier:  Cayman Chemical 336761 MG
Encompass_Preferred
A synthetic ceramide and stereoisomer of C18 ceramide; inhibits rat brain mt-CDase (IC50 = 16.6 µM)
Catalog No. 50-294-7769