Learn More
Medchemexpress LLC C18 dihydroceramide | 2304-80-5 | MFCD01076443 | 99.0% | 567.97 g/mol | C36H73NO3 | 5 MG

Supplier: Medchemexpress LLC HY1415805MG
C18 dihydroceramide is a saturated dihydroceramide (sphingolipid) used as a biochemical reagent and analytical standard in lipid research. The compound (CAS 2304-80-5) is supplied as a solid for use in biochemical assays, sphingolipid metabolism studies, and analytical method development. Storage recommendations: sealed at -20°C; in solvent, store at -80°C (up to 6 months) or -20°C (up to 1 month).
- High purity suitable for analytical and research applications.
- Molecular weight 567.97 g/mol.
- Molecular formula C36H73NO3.
- Provided as a 5 mg solid quantity.
- Storage conditions support long-term stability when frozen.
- Appropriate for sphingolipid metabolism and analytical method development.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.