Learn More
Cayman Chemical C18 PhytoceramIde-d3t18 01 1mg
SDP
Supplier:  Cayman Chemical 271991 MG
Encompass_Preferred
An internal standard for the quantification of C18 phytoceramide by GC- or LC-MS
Catalog No. 50-293-7031