Learn More
Medchemexpress LLC C1s-A subcomponent protein | >85.0% | 20 UG

Supplier: Medchemexpress LLC HYP7546320UG
Recombinant mouse C1s-A subcomponent is a serine protease produced in HEK293 cells and supplied as a 0.22 μm filtered PBS solution with a C-terminal 10xHis tag. It is intended for research on the classical complement pathway, proteolytic processing, and biochemical or immunological assays. Molecular weight ≈ 76.6 kDa; purity >85% by reducing SDS-PAGE.
- Recombinant protein produced in HEK293 cells for mammalian post-translational modifications.
- C-terminal His tag facilitates affinity purification and detection.
- Supplied as a filtered PBS solution (pH 7.4) ready for dilution and use.
- High purity (>85%) as determined by reducing SDS-PAGE.
- Available in small pack sizes, including 20 μg, for pilot experiments.
- Stable when stored at -80°C; avoid repeated freeze-thaw cycles.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.