Learn More
Aviva Systems Biology Corp C20orf144 Antibody - middle region (ARP52527_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52527_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C20orf144. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C20orf144. Alternate names: dJ63M2.6ApplicationWB. Species reactivity: Human
Catalog No. 50-014-54005