Learn More
Cayman Chemical C24 1 1DeoxyceramIdem18 1500ug
SDP
Supplier:  Cayman Chemical 27563500 UG
Encompass_Preferred
A very long-chain atypical ceramide; contains a 1-deoxysphingosine (m18:1(14Z)) backbone with a cis double bond at the 14-15 position
Catalog No. 50-293-7321