Learn More
Cayman Chemical C24 1 CeramIde-d7d18 1-d72 1mg
SDP
Supplier:  Cayman Chemical 227901 MG
Encompass_Preferred
An internal standard for the quantification of C24:1 Ceramide by GC- or LC-MS
Catalog No. 50-299-5512