Learn More
Cayman Chemical C24 1-DeoxyceramIdem18 124 1mg
SDP
Supplier:  Cayman Chemical 270161 MG
Encompass_Preferred
A very long-chain atypical ceramide; has been found as a major deoxyceramide species in MEFs following application of 1-deoxysphinganine alkyne or 1-deoxysphinganine-d3
Catalog No. 50-293-8307