Learn More
Cayman Chemical C24 3sulfo GalactosylcRamI 1mg
SDP
Supplier:  Cayman Chemical 243511 MG
Encompass_Preferred
An endogenous sulfatide; the major sulfatide in mature myelin in the central and peripheral nervous systems; elevated levels are found in plasma derived from patients with metachromatic leukodystrophy
Catalog No. 50-293-3565