Learn More
Selleck Chemical LLC C25-140 5mg 1358099-18-9
SDP
Supplier:  Selleck Chemical LLC S66795MG
Encompass_Preferred
C25-140 is a small-molecule inhibitor of TRAF6-Ubc13 interaction. C25-140 directly binds to TRAF6, thereby blocking the interaction of TRAF6 with Ubc13, and as a consequence lowers TRAF6 activity. C25-140 impedes NF-?B activation in various immune and inflammatory signaling pathways also in primary human and murine cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1822731