Learn More
Cayman Chemical C26 SphngomyelIn-d4d18 12 1mg
SDP
Supplier:  Cayman Chemical 90034601 MG
Encompass_Preferred
An internal standard for the quantification of C26 sphingomyelin by GC- or LC-MS
Catalog No. 50-289-5399