Learn More
Medchemexpress LLC HY-100461 1mg Medchemexpress, C29 CAS:363600-92-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1004611MG
Encompass_Preferred
Medchemexpress, HY-100461 1mg C29 CAS:363600-92-4 C29 is a Toll-like receptor 2 (TLR2) inhibitor. C29 blocks hTLR2/1 and hTLR2/6 signaling with IC50s of 19.7 and 37.6 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8620