Learn More
TARGETMOL CHEMICALS INC C29 5mg
SDP
Supplier:  TARGETMOL CHEMICALS INC T35025MG
Encompass_Preferred
C29 is a novel inhibitor of TLR2/1 and TLR2/6 signaling induced by synthetic and bacterial TLR2 agonists in human HEK-TLR2 and THP-1 cells, but only TLR2/1 signaling in murine macrophages.
Catalog No. NC1624952