Learn More
Complement Technology Inc C3 Protein, 250 μg/vial, Frozen liquid, >95% purity, 1.0 mg/mL concentration, 10 mM sodium phosphate, 145 mM NaCl, pH 7.2, No preservative, 0.22 μm filtered
Supplier: Complement Technology Inc A113
Complement Technology Inc. Human C3 Protein is a highly purified, native human complement component 3 supplied as a frozen liquid for scientific research applications. This critical protein plays a central role in complement activation, leading to the generation of enzyme complexes that cleave C3 into active fragments. C3b is essential for opsonization and activation of further complement components. This product is ideal for researchers focusing on immune response mechanisms.
- Product name: C3 Protein
- Size: 250 μg/vial
- Concentration: 1.0 mg/mL
- Form: Frozen liquid
- Purity: >95% by SDS-PAGE
- Activity: >70% versus normal human serum standard
- Buffer: 10 mM sodium phosphate, 145 mM NaCl, pH 7.2
- Molecular weight: 185,000 Da
- Storage conditions: -70 °C or below
- Source: Normal human serum
- Glycosylation: ~2.7%
- Filtration: 0.22 μm filtered
- Origin: Manufactured in the USA
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.