Learn More
Cayman Chemical C3 BODIPY Ganglioside GM1 d1
SDP
Supplier:  Cayman Chemical 33799100 UG
Encompass_Preferred
A fluorescently labeled form of ganglioside GM1; ex/em = 503/512 nm, respectively
Catalog No. 50-294-7730