Learn More
Strem, An Ascensus Company CAS# 14871-41-1. Chlorocarbonylbis(triphenylphosphine)iridium(I), 99% VASKA'S COMPLEX MFCD00009590. 1G
SDP
Supplier:  Strem, An Ascensus Company 7703001G
Encompass
CAS# 14871-41-1. 1G. Chlorocarbonylbis(triphenylphosphine)iridium(I), 99% VASKA'S COMPLEX. MFCD00009590. Molecular Formula: C37H30ClIrOP2 Molecular Weight: 780.27. Color/Form: lemon yellow xtl. Strem#: 77-0300 www.strem.com/catalog/v3/77-0300/
Catalog No. 50-195-9585