Learn More
ATCC C3H/10T1/2 Clone 8; Embryonic Fibroblast; Mouse (Mus musculus)
Supplier:  ATCC CCL226
Encompass
C3H/10T1/2, Clone 8 is a cell line exhibiting fibroblast morphology that was isolated from a line of C3H mouse embryo cells. This cell line was deposited by C Heidelberger.
Catalog No. 50-238-4152