Learn More
Waters Corp Waters Corporation Column, C4, 300Å, 2.5 μm, 2.1 mm, 50 mm
Supplier:  Waters Corp 186009127
Encompass_Preferred
This column is optimized and QC tested for protein separations, featuring a 300Å pore size BEH particle that offers a wide usable pH range, superior low pH stability, and ultra-low column bleed.
  • Optimized and QC tested for protein separations.
  • Offers wide usable pH range, superior low pH stability, and ultra-low column bleed.
  • Provides excellent peptide separation with thin, symmetrical peaks.
  • Ensures excellent peak shape and retention in formic and trifluoroacetic acid.
  • QC verified with a peptide map for consistent proteomics and protein characterization results.
  • BEH technology provides quality performance for large biological macromolecules.
  • Manufactured in a cGMP, ISO 9002 certified facility with chromatographically tested batches.
Catalog No. NC1646812