Learn More
Cell Biologics C57BL/6 MOUSE PRIMARY PROXIMAL
SDP
Supplier:  Cell Biologics C576015
Encompass_Preferred
C57BL/6 Mouse Primary Proximal Tubular Epithelial CellsCatalog No. C57-6015Suggested MediumM6621 Complete Epithelial Cell Medium w/ Kit – 500 mlProduct DescriptionC57BL/6 Mouse Primary Proximal Tubular Epithelial Cells from Cell Biologics are isolated from the proximal tubular tissues of pathogen-free laboratory mice and grown in gelatin pre-coated tissue culture flask with Cell Biologics’ Complete Growth Medium. Cells at passage 3 are harvested from flasks and cryo-preserved in vials. Each vial contains 0.5x106 cells per ml and is delivered frozen. C57BL/6 Mouse Primary Proximal Tubular Epithelial Cells are characterized by immunofluorescent staining with E-cadherin (Catalog 610182, BD) or ZO-1 (Catalog 617300, Life Technology) antibody. These cells are negative for bacteria, yeast, fungi, and mycoplasma and can be expanded for 3-5 passages at a split ratio of 1:2 under the cell culture conditions specified by Cell Biologics.
Catalog No. NC2391705