Learn More
Gold Biotechnology Inc C58C1 pSuperAgro v4 Agro Comp
SDP
Supplier:  Gold Biotechnology Inc CC55410X50
Encompass_Preferred
GoldBio s C58C1 Chemically Competent Agrobacterium cells allow you to obtain high transformation efficiency in applications such as gDNA or cDNA library construction C58C1 has the chromosomal backbone of C58 but it has been cured of the Ti plasmid pTiC58 Our C58C1 strain harbors streptomycin and rifampicin resistance genes C58C1 Competent cells are optimized for genetic transformation of plants such as Arabidopsis but can be used in many other plants
Catalog No. 50-005-62342