Learn More
Enzo Life Sciences C5a receptor antagonist W-54011 (5mg). CAS: 405098-33-1
SDP
Supplier:  Enzo Life Sciences ENZCHM1220005
Encompass_Preferred
Alternative name: N-((4-Dimethylaminophenyl)methyl)-N-(4-isopropylphenyl)-7-methoxy-1,2,3,4-tetrahydronaphthalen-1-carboxamide . HCl. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO or methanol. Long Term Storage: -80°C.
Catalog No. 50-201-3408