Learn More
Abcam C6 Ceramide, biologically active ceramide analog, 5MG
Supplier:  Abcam AB1417435MG
Encompass_Preferred
MW 397.6 Da, Purity >98%. Non-physiological and biologically active ceramide analog. Various pharmacological activities including activation of protein phosphatase 2A and MAPK, suppression of insulin-induced tyrosine phosphorylation and induction of cell cycle arrest at the G0/G1 transition. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6454