Learn More
Cayman Chemical C6 CRamIDd18 16 0 Alkyn 100ug
SDP
Supplier:  Cayman Chemical 24519100 UG
Encompass_Preferred
An ω-alkyne derivative of C6 ceramide; can be used in click chemistry reactions to tag C6 ceramide with fluorescent or biotinylated labels for analysis of its metabolism and biological activity
Catalog No. 50-301-5843