Learn More
Cayman Chemical C6 dIhydro CeramIded18 065mg
SDP
Supplier:  Cayman Chemical 243685 MG
Encompass_Preferred
A metabolically inactive analog of C6 ceramide; has been used as a negative control in experiments investigating the biological activity of C6 ceramide
Catalog No. 50-293-3861