Learn More
Selleck Chemical LLC C646 10mg 328968-36-1
SDP
Supplier:  Selleck Chemical LLC S715210MG
Encompass_Preferred
C646 is an inhibitor for histone acetyltransferase, and inhibits p300 with a Ki of 400 nM in a cell-free assay. Preferentially selective for p300 versus other acetyltransferases. C646 induces cell cycle arrest, apoptosis and autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1001454