Learn More
Supply Solutions C7 - C40 Saturated Alkanes Standard, 1mL ampule
Supplier:  Supply Solutions SIGMA 49452U
Encompass_Preferred
Certified reference material, 1000 ?g/mL each component in hexane
Catalog No. NC0829462