Learn More
Medchemexpress LLC C7/Complement compon 5ug
SDP
Supplier:  Medchemexpress LLC HYP743715UG
Encompass_Preferred
C7 also known as complement component C7 protein is a key component of the membrane attack complex (MAC) and plays a key role in innate and adaptive immune responses As a component of MAC C7 actively participates in the formation of target cell plasma membrane pores C7/Complement component C7 Protein Human (HEK293 His) is the recombinant human-derived C7/Complement component C7 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-78940