Learn More
AdipoGen C9-L-HSL
SDP
Supplier:  AdipoGen CDXN0236M050
Encompass_Preferred
Chemical. CAS 177158-21-3. Formula C13H23NO3. MW 241.33. N-Nonanoyl-L-homoserine lactone is a small diffusible signaling molecule involved in quorum sensing, controlling gene expression and affecting cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence in general, infection prevention, and formation of biofilms.
Catalog No. 50-256-0388