Learn More
Enzo Life Sciences CA-074 (1mg). CAS: 134448-10-5
SDP
Supplier:  Enzo Life Sciences ALX260017M001
Encompass_Preferred
Cathepsin B inhibitor. Potent and specific inhibitor of cathepsin B in vitro and in vivo. Free acid form used in vitro or in isolated enzyme studies but not in whole cells. Can discriminate between cathepsin B and L/S forms. Alternative name: N-[L-3-trans-Propylcarbamoyloxirane-2-carbonyl]-Ile-Pro-OH. Formula: C18H29N3O6. MW: 383.5. Purity: ≥98% (HPLC). Solubility: Soluble in ethyl acetate (50mg/ml) or DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8224