Learn More
Medchemexpress LLC CA-170 10mg
SDP
Supplier:  Medchemexpress LLC HY10109310MG
Encompass_Preferred
CA-170 is an orally delivered dual inhibitor of VISTA and PD-L1 CA-170 exhibits potent rescue of proliferation and effector functions of T cells inhibited by PD-L1/L2 and VISTA with selectivity over other immune checkpoint proteins as well as a broad panel of receptors and enzymes1]2]
Catalog No. 50-002-11836