Learn More
Selleck Chemical LLC CA-170
SDP
Supplier:  Selleck Chemical LLC S847325MG
Encompass_Preferred
CA-170 (AUPM-170 PD-1-IN-1 ) is potent and orally available inhibitor of the immune checkpoint regulatory proteins PD-L1 (programmed cell death ligand-1) and VISTA (V-domain immunoglobulin (Ig) suppressor of T-cell activation) CA-170 exhibits anti-tumor efficacy
Catalog No. 50-000-01716