Learn More
Enzo Life Sciences Carbonic anhydrase 2 (human), (recombinant) (10µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT1660010
Encompass_Preferred
Alternative name: CA2, Carbonic anhydrase II, CA-II, Car2. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized from 0.22µm filtered solution in 20mM TRIS, pH 8.0, containing 150mM NaCl and 1mM DTT. Normally mannitol or trehalose is added as protectant before lyophilization. Source: Produced in HEK 293 cells. Long Term Storage: -20°C.
Catalog No. 50-201-3954