Learn More
Medchemexpress LLC Ca inhibitor 1 (GS-6207 analog) | 2189684-45-3 | 99.8% | 958.34 g·mol^-1 | C41H36ClF8N7O5S2 | 10MM 1ML

Supplier: Medchemexpress LLC HY12459410MM 1ML
CA inhibitor 1 is a synthetic HIV capsid inhibitor and alkyne-containing small molecule supplied as a ready-to-use 10 mM solution in DMSO for research and chemical biology applications, including copper-catalyzed azide-alkyne cycloaddition workflows.
- Supplied as 10 mM solution in DMSO
- High purity: 99.8%
- Molecular weight 958.34 g·mol⁻¹
- Molecular formula C41H36ClF8N7O5S2
- Contains terminal alkyne for click chemistry applications
- Recommended storage: powder -20 °C; in solvent -80 °C for long-term storage
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.