Learn More
ATCC Ca Ski; Cervical Carcinoma; Human (Homo sapiens)
Supplier:  ATCC CRL1550
Encompass
Ca Ski is a cell line exhibiting epithelial morphology that was isolated from the cervix of a White, 40-year-old, female patient with epidermoid carcinoma. This cell line was deposited by RA Pattillo and can be used in cancer, infectious disease, and sexually transmitted disease research.
Catalog No. 50-238-2478