Learn More
Medchemexpress LLC CA12/Carbonic Anhydr 50ug

Supplier: Medchemexpress LLC HYP7172050UG
Recombinant human carbonic anhydrase XII (CA12) expressed in Pichia pastoris and purified with an N-terminal 6xHis tag. The lyophilized protein corresponds to UniProt O43570 (A25-S301), is >90% pure by reducing SDS-PAGE, and is supplied with formulation and storage recommendations for reliable use in biochemical assays and antibody development. For research use only.
- Recombinant human protein expressed in Pichia pastoris.
- N-terminal 6xHis tag for easy purification and detection.
- Sequence covers A25-S301 of UniProt O43570.
- Purity greater than 90% as determined by reducing SDS-PAGE.
- Lyophilized formulation for extended shelf life and transport.
- Stable storage recommendations provided for short and long term.
- Suitable for biochemical assays, antibody development, and reference use.
- Intended for research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.