Learn More
Genscript Corporation CA125 C33 mAb Mouse
Supplier:  Genscript Corporation V059011
Encompass
CA125 known as cancer antigen 125 carcinoma antigen 125 and carbohydrate antigen 125 is a member of the mucin family glycoproteins It is involved in boosting tumorigenesis and tumor proliferation and is considered as a useful biomarker for the diagnosis of ovarian cancer
Catalog No. 50-000-57325