Learn More
Genscript Corporation CA125 D34 mAb Mouse
Supplier:  Genscript Corporation V05902100
Encompass
CA125 known as cancer antigen 125 carcinoma antigen 125 and carbohydrate antigen 125 is a member of the mucin family glycoproteins It is involved in boosting tumorigenesis and tumor proliferation and is considered as a useful biomarker for the diagnosis of ovarian cancer
Catalog No. 50-000-57734
Only null left
Add to Cart
Request a Quote